Assay Details
Gene Symbol: MKRN7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026640.1 | 1 | 549 | 73 |
Gene Symbol: ZNF334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017027944.1 | XP_016883433.1 | 10 | 4684 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
zinc finger protein 663, pseudogene |
ZNF663P | ZNF663 | -6.0kb | Sense | Chr. 20 - 46456493 - 46457643 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000902.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P209624 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4687 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4686 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 43212 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38494 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P209624 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3333985 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P209624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191451 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191451 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195724 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191422 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191413 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191446 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191457 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191436 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191427 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191432 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191450 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195699 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191401 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191415 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191419 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191412 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 217115_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR20012415.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20012415.hg.1 | TC20000902.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009272.hg.1 | ENST00000433469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164988.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164988.hg.1 | TC2000009272.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164988.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164990.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164990.hg.1 | TC2000009272.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164990.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009272.hg.1 | ENST00000433469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009272.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164989.hg.1 | ENST00000433469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164989.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164989.hg.1 | TC2000009272.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164989.hg.1 | ENST00000433469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164989.hg.1 | NR_026640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007018.hg.1 | ENST00000444706.2 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017745 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017744 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017745 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017744 | - | - |
