Assay Details
| Gene Symbol: LOC100240735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026658.1 | 1 - 2 | 186 | 78 | ||
| GenBank mRNA | CN361140.1 | - | 1 - 2 | 186 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC440101 | FLJ12825 | Anti-sense | Chr. 12 - 54058254 - 54122234 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC100240734 | LOC100240734 | 20.4kb | Anti-sense | Chr. 12 - 54102209 - 54102699 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001561.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P170481 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4701 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4700 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 21969 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8292 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47676 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P170481 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P170481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798144 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08363027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798144 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28798105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08363027 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08362980 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16560553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08362982 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08362977 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08362978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010139.hg.1 | ENST00000611630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010139.hg.1 | ENST00000611630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020643.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020643.hg.1 | TC12001561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010140.hg.1 | ENST00000613427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010140.hg.1 | ENST00000613427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010189.hg.1 | ENST00000377547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010189.hg.1 | ENST00000377547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020644.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020644.hg.1 | TC12001561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020645.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020645.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020645.hg.1 | TC12001561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008786.hg.1 | ENST00000454331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008786.hg.1 | ENST00000454331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020642.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020642.hg.1 | TC12001561.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183037.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010189.hg.1 | ENST00000377547.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010189.hg.1 | ENST00000377547.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183032.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183032.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183032.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183043.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183036.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000514503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010827.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008786.hg.1 | ENST00000454331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008786.hg.1 | ENST00000454331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000510754.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183035.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183031.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183031.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183031.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183033.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | ENST00000607809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183034.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200183042.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183042.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183042.hg.1 | ENST00000505259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183042.hg.1 | NR_026658.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183042.hg.1 | TC1200010827.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010140.hg.1 | ENST00000613427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010140.hg.1 | ENST00000613427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000377548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007327.hg.1 | ENST00000448436.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010139.hg.1 | ENST00000611630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010139.hg.1 | ENST00000611630.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047649 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047649 | - | - |

LOC100240735
Genomic Map