Assay Details
| Gene Symbol: FOXO3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026718.1 | 1 | 3254 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 286B | ZNF286B |
ZNF286C, ZNF286L, ZNF590 |
Sense | Chr. 17 - 18658429 - 18682259 on NCBI Build GRCh38 | |
| Downstream | tripartite motif containing 16-like | TRIM16L | TRIM70 | 24.8kb | Anti-sense | Chr. 17 - 18697998 - 18736119 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16953739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30815535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10259464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30815594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10259467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16953798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16953736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30815523 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4735 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4734 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P118418 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P118418 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240392 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P102062 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375665 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00806684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16953798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10259467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30815568 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30815594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30815568 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30815594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16953798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10259467 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P118418 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P118418 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03223826 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 119258 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 102561 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40772521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10259466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8013308 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8013307 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000395675.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000395675.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000578214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000285274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000395675.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000395675.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000442380.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000443457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000454745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000545289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000578214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000580145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | ENST00000583002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NM_001145045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012371.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205629.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205629.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205629.hg.1 | TC1700012371.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205630.hg.1 | ENST00000578214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205630.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205630.hg.1 | ENST00000578214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205630.hg.1 | NR_026718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205630.hg.1 | TC1700012371.hg.1 | Clariom D TC |
| AffymetrixPathwork Diagnostics Pathchip | pw547708 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210655_s_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008035 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008035 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 194114 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1844692 | NR_026718.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1844692 | BC015367.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1844692 | BC045800.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1844692 | AK024103.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1844692 | NM_001455.3 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7380041 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_102192 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844692 | NR_026718.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844692 | BC015367.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844692 | BC045800.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844692 | AK024103.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844692 | NM_001455.3 | human_transcriptome |

FOXO3B
Genomic Map