Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010109.hg.1 |
TC03000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010109.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010109.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010110.hg.1 |
TC03000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010110.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010110.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159031.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159031.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159031.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159031.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159031.hg.1 |
TC0300008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300159030.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159030.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159030.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159030.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159030.hg.1 |
TC0300008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
ENST00000494582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008295.hg.1 |
NR_026767.1 |
Refseq |