Assay Details
| Gene Symbol: PSORS1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026816.1 | 2 - 3 | 291 | 81 | ||
| GenBank mRNA | AY484516.1 | - | 2 - 3 | 291 | 81 | |
| LC027945.1 | - | 3 - 4 | 583 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | POU class 5 homeobox 1 | POU5F1 |
OCT3, OCT4, OTF-3, OTF3, OTF4, Oct-3, Oct-4 |
-3.0kb | Sense | Chr. 6 - 31164337 - 31170693 on NCBI Build GRCh38 |
| Downstream | HLA complex group 27 (non-protein coding) | HCG27 |
bCX101P6.9, bPG299F13.9, bQB115I13.2 |
23.5kb | Anti-sense | Chr. 6 - 31197760 - 31203968 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001522.hg.1, TC6_cox_hap2000154.hg.1, TC6_dbb_hap3000143.hg.1, TC6_mann_hap4000131.hg.1, TC6_mcf_hap5000131.hg.1, TC6_qbl_hap6000144.hg.1, TC6_ssto_hap7000126.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 317344 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 400569 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 121259 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AB020699.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AK001349.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AK025896.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AK125914.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AK126227.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AK128029.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | AL110250.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | BC010222.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | BC063863.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170092 | NM_015329.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 3349 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3297020 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4793 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4794 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50155 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3297020 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3297020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834410 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834410 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642843 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642847 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642838 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642821 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642832 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642759 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642795 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642785 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642812 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642851 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642807 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642834 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642846 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15799816 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642768 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642757 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642811 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642819 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642828 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642777 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642817 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642805 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642769 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642809 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642764 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642830 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642849 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642836 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642803 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8179729 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124899 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8179730 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8179728 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124900 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124898 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8124897 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8179727 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 238997_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11744971_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11744971_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002599.hg.1 | TC6_dbb_hap3000143.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002599.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017298.hg.1 | TC06001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017298.hg.1 | ENST00000412143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017298.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002330.hg.1 | TC6_mcf_hap5000131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002330.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002182.hg.1 | TC6_mann_hap4000131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002182.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002663.hg.1 | TC6_cox_hap2000154.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002663.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002639.hg.1 | TC6_qbl_hap6000144.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002639.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002323.hg.1 | TC6_ssto_hap7000126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002323.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177031.hg.1 | ENST00000412143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177031.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177031.hg.1 | ENST00000412143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177031.hg.1 | NR_026816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177031.hg.1 | TC0600011407.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 238997_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 238997_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238997_at2 | - | - |
| Human Genome U95 Set | 86474_at | - | - |
| Human Genome U95 Set | 86474_PM_at | - | - |
| Human Genome U133 Set | 238997_at | - | - |
| Human Genome U133 Set | 238997_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029687 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029687 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143712 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309634 | NR_026816.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3309634 | AY484516.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309634 | NR_026816.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309634 | AY484516.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309634 | NR_026816.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309634 | AY484516.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309634 | NR_026816.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309634 | AY484516.1 | human_transcriptome |

PSORS1C3
Genomic Map