| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038089.hg.1 |
TC01002465.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038089.hg.1 |
ENST00000441044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038089.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038088.hg.1 |
TC01002465.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038088.hg.1 |
ENST00000441044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038088.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038090.hg.1 |
TC01002465.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038090.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201153.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201163.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201163.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201163.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201163.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201163.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201152.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201150.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201159.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201164.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201164.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201164.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000403496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000423995.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013603.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201160.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201160.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201160.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201160.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201160.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
ENST00000426597.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
ENST00000441044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
NR_026850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201162.hg.1 |
TC0100013603.hg.1 |
Clariom D TC |