| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007538.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007538.hg.1 |
TC19000552.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007537.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007537.hg.1 |
TC19000552.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007539.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007539.hg.1 |
TC19000552.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900160886.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160886.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160886.hg.1 |
TC1900008073.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160886.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160886.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160893.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160893.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160893.hg.1 |
TC1900008073.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160893.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160893.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
TC1900008073.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160890.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008073.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
NR_026870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
TC1900008073.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000594263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000597077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
ENST00000597791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160891.hg.1 |
NR_026870.1 |
Refseq |