Assay Details
| Gene Symbol: H1FX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026991.1 | 1 - 2 | 1075 | 61 | ||
| GenBank mRNA | AL353963.1 | - | 1 - 2 | 367 | 61 | |
| BM709882.1 | - | 1 - 2 | 301 | 61 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | ENST00000626358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | ENST00000333762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | ENST00000626358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012384.hg.1 | NM_006026.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000383461.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000433902.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000502789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000511998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | ENST00000537780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008770.hg.1 | NR_026991.1 | Refseq |

H1FX-AS1
Genomic Map