Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014415.hg.1 |
TC06001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014415.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014415.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014415.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199998.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000434292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
ENST00000612554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014212.hg.1 |
NR_027116.2 |
Refseq |