Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003953.hg.1 |
TC07000258.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003953.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003953.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150720.hg.1 |
TC0700007312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000415848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000420821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
ENST00000422822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007312.hg.1 |
NR_027119.1 |
Refseq |