Assay Details
| Gene Symbol: AATK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027255.1 | 1 - 2 | 242 | 81 | ||
| GenBank mRNA | AK126811.1 | - | 1 - 2 | 242 | 81 | |
| BC140737.1 | - | 1 - 2 | 233 | 81 | ||
| BC171777.1 | - | 1 - 2 | 233 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1250 | MIR1250 |
MIRN1250, hsa-mir-1250, mir-1250 |
-32.2kb | Anti-sense | Chr. 17 - 81133196 - 81133308 on NCBI Build GRCh38 |
| Downstream | centrosomal protein 131 | CEP131 |
AZ1, AZI1, ZA1 |
22.7kb | Anti-sense | Chr. 17 - 81189593 - 81222989 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

AATK-AS1
Genomic Map