| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013343.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013343.hg.1 |
TC19000972.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013342.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013342.hg.1 |
TC19000972.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
TC1900011831.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203343.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
TC1900011831.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203345.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
TC1900011831.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203344.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000593642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
ENST00000600726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011831.hg.1 |
NR_027334.2 |
Refseq |