Assay Details
| Gene Symbol: URAHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027335.2 | 1 - 2 | 94 | 76 | ||
| GenBank mRNA | BC044232.1 | - | 1 - 2 | 94 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth arrest specific 8 | GAS8 |
CILD33, DRC4, GAS11 |
-2.0kb | Anti-sense | Chr. 16 - 90019629 - 90044971 on NCBI Build GRCh38 |
| Downstream | PR domain 7 | PRDM7 |
PFM4, ZNF910 |
8.9kb | Sense | Chr. 16 - 90056566 - 90077339 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001363.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16018879.hg.1 | ENST00000409873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018879.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018879.hg.1 | TC16001363.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018877.hg.1 | ENST00000409873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018877.hg.1 | ENST00000517889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018877.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018877.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018877.hg.1 | TC16001363.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600194928.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194928.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194928.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194928.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194928.hg.1 | TC1600011586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409768.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011586.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194927.hg.1 | TC1600011586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000409873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000517889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000521551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | ENST00000610227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | NR_027335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | NR_027336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194925.hg.1 | TC1600011586.hg.1 | Clariom D TC |

URAHP
Genomic Map