Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027353.1 | 1 - 2 | 202 | 108 | ||
| GenBank mRNA | AK300089.1 | - | 1 - 2 | 168 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | required for meiotic nuclear division 5 homolog A | RMND5A |
CTLH, GID2, GID2A, RMD5, p44CTLH |
-12.7kb | Anti-sense | Chr. 2 - 86720291 - 86778041 on NCBI Build GRCh38 |
| Downstream | CD8b molecule | CD8B |
CD8B1, LEU2, LY3, LYT3, P37 |
23.6kb | Sense | Chr. 2 - 86815337 - 86861924 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300089.1 transcripts.
Set Membership: |
|

CD8A
Genomic Map