Assay Details
| Gene Symbol: UG0898H09 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027378.1 | 1 | 4949 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929628 | LOC101929628 | -925.5kb | Sense | Chr. 8 - 61785072 - 62100736 on NCBI Build GRCh38 | |
| Downstream | gamma-glutamyl hydrolase | GGH | GH | 30.2kb | Anti-sense | Chr. 8 - 63015079 - 63039051 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3378614 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5136 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5137 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P12552 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41094 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 40485 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42022 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3378614 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3378614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450610 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450610 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06167497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26450618 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P170275 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P12552 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P170275 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P12552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167534 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16100562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167536 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16100564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167482 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167493 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167525 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167475 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167502 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16100566 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06167537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8146634 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8146633 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1558388_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1558387_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000622794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000622794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153671.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153671.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153671.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153671.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153671.hg.1 | TC0800007803.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000622794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000622794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | ENST00000623646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007803.hg.1 | NR_027378.1 | Refseq |
| Human Genome U95 Set | 57981_PM_at | - | - |
| Human Genome U95 Set | 57981_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036649 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036649 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1697476 | NR_027378.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1697476 | NR_027378.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1697476 | NR_027378.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1697476 | NR_027378.1 | human_transcriptome |

UG0898H09
Genomic Map