Assay Details
| Gene Symbol: LOC100129034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027406.1 | 1 | 640 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myeloproliferative disease associated tumor antigen 5 | LOC613206 | -92.2kb | Anti-sense | Chr. 9 - 124258585 - 124261417 on NCBI Build GRCh38 | |
| Downstream | adhesion G protein-coupled receptor D2 | ADGRD2 |
GPR144, PGR24 |
91.9kb | Sense | Chr. 9 - 124451087 - 124477795 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000637.hg.1
Set Membership: |
|

LOC100129034
Genomic Map