| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010396.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010396.hg.1 |
TC17000799.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000613178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010397.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010397.hg.1 |
TC17000799.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010394.hg.1 |
ENST00000435469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010394.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010394.hg.1 |
TC17000799.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165112.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165112.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165112.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165118.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165118.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165118.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165103.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165122.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165122.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165122.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165121.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165121.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165121.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000435469.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000590734.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000613178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
ENST00000622750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008721.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165111.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165111.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165111.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165124.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165124.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165124.hg.1 |
ENST00000620266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700165124.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165124.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165100.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165100.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165100.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700165123.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165123.hg.1 |
NR_027418.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700165123.hg.1 |
TC1700008721.hg.1 |
Clariom D TC |