Assay Details
| Gene Symbol: LOC100128164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024409.2 | 1 - 2 | 758 | 69 | ||
| GenBank mRNA | AK098335.1 | - | 1 - 2 | 758 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 7 | SAMD7 | -15.1kb | Anti-sense | Chr. 3 - 169911572 - 169939175 on NCBI Build GRCh38 | |
| Downstream |
SEC62 homolog, preprotein translocation factor |
SEC62 |
Dtrp1, HTP1, TLOC1, TP-1 |
58.0 | Anti-sense | Chr. 3 - 169966792 - 169998373 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001986.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03035551.hg.1 | TC03001986.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035551.hg.1 | ENST00000483289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035551.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035551.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035553.hg.1 | TC03001986.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035553.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035553.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000494834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035550.hg.1 | TC03001986.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035550.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197861.hg.1 | TC0300013086.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197860.hg.1 | TC0300013086.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197863.hg.1 | TC0300013086.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300197858.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197858.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197858.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197858.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197858.hg.1 | TC0300013086.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197859.hg.1 | TC0300013086.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000469301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000483289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000487580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000494834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | ENST00000600502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_024409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013086.hg.1 | NR_027622.1 | Refseq |

LOC100128164
Genomic Map