Assay Details
| Gene Symbol: CLCA3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024604.1 | 11 - 12 | 1766 | 75 | ||
| GenBank mRNA | AF043976.1 | - | 11 - 12 | 1767 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride channel accessory 4 | CLCA4 |
CaCC, CaCC2 |
-67.0kb | Sense | Chr. 1 - 86547076 - 86580758 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105378828 | LOC105378828 | 54.9kb | Anti-sense | Chr. 1 - 86571181 - 86704493 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CLCA3P
Genomic Map