Assay Details
| Gene Symbol: XRCC6P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024608.1 | 7 - 8 | 855 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | endogenous retrovirus group FC1 Env polyprotein | LOC105373297 |
ERVFC1, Fc1env |
-1.9mb | Anti-sense | Chr. X - 97841749 - 97849381 on NCBI Build GRCh38 |
| Downstream | protocadherin 19 | PCDH19 |
EFMR, EIEE9 |
570.6kb | Sense | Chr. X - 100291644 - 100410273 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001206.hg.1
Set Membership: |
|

XRCC6P5
Genomic Map