Assay Details
| Gene Symbol: LOC389033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026740.1 | 3 | 349 | 68 | ||
| GenBank mRNA | AI161103.1 | - | 1 | 68 | ||
| BF592795.1 | - | 1 | 68 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC151121 | LOC151121 | -649.0kb | Sense | Chr. 2 - 129242173 - 129273891 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927924 | LOC101927924 | 178.0 | Anti-sense | Chr. 2 - 129923177 - 129946703 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002299.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 153637 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 512865 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 272062 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 791605 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 139974 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 214508 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 316678 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 103487 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 61652 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 49721 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 326965 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 347102 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 111497 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3283601 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6807 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54166 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P13397670 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00468206 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3283601 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3283601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593968 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15321933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15321952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15321933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15321952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593968 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21593982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01478188 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P230736 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478183 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478186 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321884 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478170 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478188 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321926 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478152 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321902 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478179 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478144 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321880 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15321889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478146 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478174 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478165 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478175 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01478150 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00468206 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13393244 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02036897.hg.1 | TC02002299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036897.hg.1 | ENST00000433290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036897.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036896.hg.1 | TC02002299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036896.hg.1 | ENST00000433290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036896.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036895.hg.1 | TC02002299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036895.hg.1 | ENST00000433290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036895.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203931.hg.1 | TC0200014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203933.hg.1 | TC0200014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203939.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203939.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203939.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203939.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203939.hg.1 | TC0200014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000397540.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000433290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | ENST00000440031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014240.hg.1 | NR_026740.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014325 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014325 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1679632 | NR_026740.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1679632 | AI161103.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679632 | NR_026740.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679632 | AI161103.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1679632 | NR_026740.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1679632 | AI161103.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1679632 | NR_026740.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1679632 | AI161103.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1679632 | NR_026740.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1679632 | AI161103.1 | human_transcriptome |

LOC389033
Genomic Map