Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013968.hg.1 |
TC06001171.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013968.hg.1 |
NR_027284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010168.hg.1 |
ENST00000629776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010168.hg.1 |
NR_027284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169501.hg.1 |
ENST00000629776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169501.hg.1 |
NR_027284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169501.hg.1 |
ENST00000629776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169501.hg.1 |
NR_027284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169501.hg.1 |
TC0600010168.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010168.hg.1 |
ENST00000629776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010168.hg.1 |
NR_027284.1 |
Refseq |