Assay Details
| Gene Symbol: LOC101927709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110272.1 | 1 - 2 | 391 | 106 | ||
| GenBank mRNA | AL117625.1 | - | 1 - 2 | 400 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | insulin induced gene 2 | INSIG2 | INSIG-2 | -492.3kb | Anti-sense | Chr. 2 - 118088418 - 118110031 on NCBI Build GRCh38 |
| Downstream | engrailed homeobox 1 | EN1 | 239.2kb | Sense | Chr. 2 - 118842171 - 118848183 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927709
Genomic Map