Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | D-tyrosyl-tRNA deacylase 1 | DTD1 |
C20orf88, DUE-B, DUEB, HARS2, pqn-68 |
Anti-sense | Chr. 20 - 18587893 - 18763917 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929526 | LOC101929526 | 84.5kb | Sense | Chr. 20 - 18674395 - 18698708 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map