Assay Details
| Gene Symbol: VPS13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF070595.1 | - | 1 | 653 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | G protein subunit alpha 14 | GNA14 | 1.5kb | Anti-sense | Chr. 9 - 77423079 - 77648316 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

VPS13A
Genomic Map