Assay Details
| Gene Symbol: PIAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324046.1 | NP_001310975.1 | 16 | 11209 | 94 | |
| NM_001324047.1 | NP_001310976.1 | 15 | 11149 | 94 | ||
| NM_001324048.1 | NP_001310977.1 | 16 | 11235 | 94 | ||
| NM_001324049.1 | NP_001310978.1 | 17 | 11358 | 94 | ||
| NM_004671.4 | NP_004662.2 | 14 | 11037 | 94 | ||
| NR_136684.1 | 13 | 10952 | 94 | |||
| GenBank mRNA | AF086240.1 | - | 1 | 446 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5 | ST8SIA5 |
SIAT8-E, SIAT8E, ST8SiaV |
-46.1kb | Sense | Chr. 18 - 46679118 - 46757169 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PIAS2
Genomic Map