Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 L6 | UBE2L6 |
RIG-B, UBCH8 |
-42.8kb | Sense | Chr. 11 - 57551655 - 57568330 on NCBI Build GRCh38 |
| Downstream | microRNA 130a | MIR130A |
MIRN130A, miRNA130A, mir-130a |
29.6kb | Anti-sense | Chr. 11 - 57641198 - 57641286 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map