Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADAMTS9 antisense RNA 2 | ADAMTS9-AS2 | -52.0kb | Sense | Chr. 3 - 64684870 - 65011468 on NCBI Build GRCh38 | |
| Downstream |
membrane associated guanylate kinase, WW and PDZ domain containing 1 |
MAGI1 |
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3 |
266.7kb | Anti-sense | Chr. 3 - 65353525 - 66038836 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map