Assay Details
| Gene Symbol: EDNRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF114163.1 | - | 1 | 552 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EDNRB antisense RNA 1 | EDNRB-AS1 | -10.1kb | Anti-sense | Chr. 13 - 77818937 - 77908442 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1069 | LINC01069 | 60.4kb | Sense | Chr. 13 - 77979766 - 77994773 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000334286.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000377211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000475537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | ENST00000626030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_000115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001122659.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_001201397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009309.hg.1 | NM_003991.3 | Refseq |

EDNRB
Genomic Map