Assay Details
| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712970.3 | XP_006713033.1 | 6 | 6041 | 84 | |
| XM_017005718.1 | XP_016861207.1 | 6 | 6041 | 84 | ||
| XM_017005722.1 | XP_016861211.1 | 5 | 6023 | 84 | ||
| GenBank mRNA | AF131805.1 | - | 1 | 1550 | 84 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BX115489.1 | - | 2 | Chr. 3 - 15404516 - 15406543 [+] on Build GRCh38 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3 domain binding protein 5 | SH3BP5 |
SAB, SH3BP-5 |
-64.7kb | Sense | Chr. 3 - 15254353 - 15341394 on NCBI Build GRCh38 |
| Downstream | ELL associated factor 1 | EAF1 | 19.8kb | Anti-sense | Chr. 3 - 15427557 - 15442613 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

METTL6
Genomic Map