Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EGF like repeats and discoidin domains 3 | EDIL3 | DEL1 | Sense | Chr. 5 - 83940595 - 84384867 on NCBI Build GRCh38 | |
| Downstream |
neuroblastoma breakpoint family member 22, pseudogene |
NBPF22P | 2.3mb | Anti-sense | Chr. 5 - 86282444 - 86297547 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map