Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methyltransferase like 15 | METTL15 | METT5D1 | Anti-sense | Chr. 11 - 28108251 - 28527041 on NCBI Build GRCh38 | |
| Downstream | microRNA 8068 | MIR8068 | hsa-mir-8068 | 347.3kb | Sense | Chr. 11 - 28477481 - 28477548 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map