Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and SOCS box containing 11 | ASB11 | -12.0kb | Sense | Chr. X - 15281697 - 15315624 on NCBI Build GRCh38 | |
| Downstream | c-fos induced growth factor | FIGF |
VEGF-D, VEGFD |
16.6kb | Sense | Chr. X - 15345591 - 15384413 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map