Assay Details
| Gene Symbol: SCN3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511610.2 | XP_011509912.1 | 3 | 491 | 85 | |
| XM_017004663.1 | XP_016860152.1 | 3 | 491 | 85 | ||
| GenBank mRNA | DA772987.1 | - | 3 | 491 | 85 | |
| DQ993538.1 | - | 1 | 65 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 38 member 11 | SLC38A11 | AVT2 | -224.0kb | Sense | Chr. 2 - 164897745 - 164955721 on NCBI Build GRCh38 |
| Downstream | cysteine and serine rich nuclear protein 3 | CSRNP3 |
FAM130A2, PPP1R73, TAIP-2, TAIP2 |
290.0kb | Anti-sense | Chr. 2 - 165469647 - 165689407 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011511610.2, XM_017004663.1 transcripts.
Set Membership: |
|

SCN3A
Genomic Map