Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAD18, E3 ubiquitin protein ligase |
RAD18 | RNF73 | -208.6kb | Sense | Chr. 3 - 8877196 - 8969249 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927416 | LOC101927416 | 18.4kb | Anti-sense | Chr. 3 - 9192493 - 9194453 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map