Assay Details
| Gene Symbol: USP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267175.3 | XP_005267232.1 | 14 | 4313 | 113 | |
| GenBank mRNA | AK021831.1 | - | 1 | 250 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927365 | LOC101927365 | -27.6kb | Anti-sense | Chr. 6 - 99424871 - 99431315 on NCBI Build GRCh38 | |
| Downstream | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 | TSTD3 | 60.7kb | Anti-sense | Chr. 6 - 99424854 - 99579288 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

USP45
Genomic Map