Assay Details
| Gene Symbol: LMBRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021916.1 | - | 1 | 1446 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor B3 | ADGRB3 | BAI3 | -298.9kb | Anti-sense | Chr. 6 - 68635282 - 69389511 on NCBI Build GRCh38 |
| Downstream | collagen type XIX alpha 1 chain | COL19A1 |
COL9A1L, D6S228E |
175.7kb | Anti-sense | Chr. 6 - 69866556 - 70215063 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LMBRD1
Genomic Map