Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAGI1 antisense RNA 1 | MAGI1-AS1 | Anti-sense | Chr. 3 - 65893816 - 65925297 on NCBI Build GRCh38 | ||
| Downstream | solute carrier family 25 member 26 | SLC25A26 |
COXPD28, SAMC |
220.6kb | Anti-sense | Chr. 3 - 66133610 - 66380021 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map