Assay Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 4083 | 97 | |
| GenBank mRNA | AK023059.1 | - | 1 | 1054 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7161 | MIR7161 | hsa-mir-7161 | -1.2kb | Sense | Chr. 6 - 158609707 - 158609790 on NCBI Build GRCh38 |
| Downstream | dynein light chain Tctex-type 1 | DYNLT1 |
CW-1, TCTEL1, tctex-1 |
23.2kb | Anti-sense | Chr. 6 - 158636474 - 158644786 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMEM181
Genomic Map