Assay Details
| Gene Symbol: LOC101928291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023086.1 | - | 1 | 1945 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cornichon family AMPA receptor auxiliary protein 3 | CNIH3 | CNIH-3 | Sense | Chr. 1 - 224434640 - 224740549 on NCBI Build GRCh38 | |
| Downstream | dynein axonemal heavy chain 14 | DNAH14 |
C1orf67, Dnahc14, HL-18, HL18 |
468.9kb | Sense | Chr. 1 - 224928422 - 225399294 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928291
Genomic Map