Assay Details
| Gene Symbol: CRIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014171.5 | NP_054890.1 | 5 | 5401 | 97 | |
| GenBank mRNA | AK023264.1 | - | 1 | 2748 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylinositol glycan anchor biosynthesis class F | PIGF | -9.3kb | Anti-sense | Chr. 2 - 46581274 - 46617112 on NCBI Build GRCh38 | |
| Downstream | suppressor of cytokine signaling 5 | SOCS5 |
CIS6, CISH6, Cish5, SOCS-5 |
69.0kb | Sense | Chr. 2 - 46698960 - 46762788 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CRIPT
Genomic Map