Assay Details
| Gene Symbol: LOC105374809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832916.1 | - | 1 | 450 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1291 | LINC01291 | Anti-sense | Chr. 2 - 74909131 - 74938418 on NCBI Build GRCh38 | ||
| Downstream | long intergenic non-protein coding RNA 1293 | LINC01293 | 9.8kb | Anti-sense | Chr. 2 - 74939347 - 74942670 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105374809
Genomic Map