Assay Details
| Gene Symbol: LOC102723694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833303.1 | - | 1 | 5949 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and AT-hook domain containing | ZFAT |
AITD3, ZFAT1, ZNF406 |
-79.0kb | Sense | Chr. 8 - 134477788 - 134713049 on NCBI Build GRCh38 |
| Downstream | microRNA 30b | MIR30B |
MIRN30B, mir-30b |
2.2kb | Sense | Chr. 8 - 134800520 - 134800607 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102723694
Genomic Map