Assay Details
| Gene Symbol: LOC105375172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC013942.1 | - | 1 | 1149 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | renal clear cell carcinoma-associated transcript 333 | KCCAT333 | Anti-sense | Chr. 7 - 17374917 - 17467256 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC101927630 | LOC101927630 | 28.8kb | Sense | Chr. 7 - 17463445 - 17558909 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105375172
Genomic Map