Assay Details
| Gene Symbol: LOC100509814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022565.1 | - | 1 | 544 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1163 | LINC01163 | TCONS_00017722 | -1.2mb | Anti-sense | Chr. 10 - 128285950 - 128317726 on NCBI Build GRCh38 |
| Downstream | early B-cell factor 3 | EBF3 |
COE3, EBF-3, O/E-2, OE-2 |
324.0kb | Sense | Chr. 10 - 129835232 - 129964274 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100509814
Genomic Map