| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009819.hg.1 |
ENST00000427169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009819.hg.1 |
ENST00000427169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000434245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015649.hg.1 |
ENST00000445201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009819.hg.1 |
ENST00000427169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009819.hg.1 |
ENST00000427169.1 |
Ensembl |