Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-) | POMGNT2 |
AGO61, C3orf39, GTDC2, MDDGA8 |
-234.4kb | Anti-sense | Chr. 3 - 43079229 - 43106092 on NCBI Build GRCh38 |
| Downstream | SNRK antisense RNA 1 | SNRK-AS1 | 4.8kb | Anti-sense | Chr. 3 - 43346919 - 43351962 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map