Assay Details
| Gene Symbol: FBXL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163315.2 | NP_001156787.2 | 9 | 4868 | 125 | |
| GenBank mRNA | AK172822.1 | - | 1 | 2150 | 123 | |
| BM682767.1 | - | 1 | 125 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ephrin A5 | EFNA5 |
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS |
-188.1kb | Sense | Chr. 5 - 107376889 - 107672364 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1023 | LINC01023 | HP07349 | 866.3kb | Sense | Chr. 5 - 108727825 - 108728261 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FBXL17
Genomic Map