Assay Details
| Gene Symbol: LINC00032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY116539.1 | - | 1 | 147 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TEK receptor tyrosine kinase | TEK |
CD202B, TIE-2, TIE2, VMCM, VMCM1 |
-17.1kb | Anti-sense | Chr. 9 - 27109141 - 27230178 on NCBI Build GRCh38 |
| Downstream | equatorin | EQTN |
AFAF, C9orf11, SPACA8 |
37.1kb | Sense | Chr. 9 - 27284654 - 27297139 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00032
Genomic Map