Assay Details
| Gene Symbol: LOC101928877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY494058.1 | - | 1 - 2 | 363 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | helt bHLH transcription factor | HELT |
HCM1228, HESL, Mgn, bHLHb44 |
-41.7kb | Sense | Chr. 4 - 185018841 - 185021265 on NCBI Build GRCh38 |
| Downstream | solute carrier family 25 member 4 | SLC25A4 |
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1 |
79.9kb | Sense | Chr. 4 - 185143263 - 185150384 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000506338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000509017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009514.hg.1 | ENST00000511746.1 | Ensembl |

LOC101928877
Genomic Map